missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) selenide, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1313014

 View more versions of this product

Catalog No. AA1313014



Specifications

Specifications

Antimony(III) selenide
(metals basis)
UN3077
Very slightly soluble in water
Pieces
3 to 12mm
611°C
1315-05-5
MFCD00016319
14,712
480.38
99.999%
Moisture Sensitive
25g

Chemical Identifiers

1315-05-5
MFCD00016319
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

Documents