Please login to your online account to display your discounted pricing

Antimony(III) oxide, 99+%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  213470050

 View more versions of this product

Catalog No. AC213470050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 291.5
Quantity 5g
CAS 1309-64-4
Chemical Name or Material Antimony(III) oxide
Assay Percent Range 99+%
Molecular Formula O3Sb2
MDL Number MFCD00011214
Synonym Antimony trioxide

Specifications

Chemical Name or Material Antimony(III) oxide
Assay Percent Range 99+%
Molecular Formula O3Sb2
Formula Weight 291.5
MDL Number MFCD00011214
Synonym Antimony trioxide