missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony(III) oxide, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 213470051

Catalog No. AC213470051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Antimony(III) oxide
5kg
O3Sb2
MFCD00011214
Antimony trioxide
291.42
99+%
1309-64-4
99% min. (Sb2O3)
Sb2O3
15,699
Solubility in water: 2.7mg/l (20°C). Other solubilities: slightly soluble in hot water,soluble in conc. hcl,acetic acid,sulfuric acids,,solutions of alkali hydroxides
291.42
Plastic Bottle
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 215-175-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only