missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony(III) oxide, 99.6% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 1157936

 View more versions of this product

Catalog No. AA1157936



Specifications

Specifications

Antimony(III) oxide
99.6% min (Metal basis)
MFCD00011214
Antimony trioxide
Powder
White
1309-64-4
Sb2O3
White powder
291.5
Odorless
500g

Chemical Identifiers

1309-64-4
Sb2O3
MFCD00011214
Antimony trioxide
Safety and Handling

Safety and Handling

H351

EINECSNumber : 215-175-0

Documents