Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) n-butoxide, 99.9% (metals basis), Sb(III) 36.0%

Chemical Identifiers

Chemical Name or Material Antimony(III) n-butoxide
CAS 2155-74-0
Formula Weight 341.1
MDL Number MFCD00015238
UN Number UN3082
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3349603

  SDS
alfa aesar™
3349603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 341.1
Chemical Name or Material Antimony(III) n-butoxide
Concentration Sb(III) 36.0%
TSCA TSCA
Density 1.264g/mL
Physical Form Liquid
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00015238
CAS 2155-74-0
Boiling Point 133° to 135°C (0.4mmHg)
UN Number UN3082
Solubility Information Soluble in anhydrous butanol, THF
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P273
Avoid release to the environment.
Quantity 1g
Flash Point >110° (230°F)
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Sensitivity Moisture Sensitive
Safety and Handling

Warning

GHS P Statement
P273
Avoid release to the environment.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.