missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) n-butoxide, 99+%

Preparation of coatings, as a catalyst

Manufacturer:  Alfa Aesar™ 4329504

 View more versions of this product

Catalog No. AA4329504



Specifications

Specifications

Antimony (III) n-butoxide
>110°C (230°F)
133°C to 135°C (0.4mmHg)
2155-74-0
MFCD00015238
1-butanol, antimony 3+ salt, acmc-20als1, antimony 3+ ; butan-1-olate, antimony 3+ ion tris butan-1-olate, antimony 3+ tributan-1-olate, antimony 3+ tris butan-1-olate, antimony iii butan-1-olate, antimony iii butoxide, antimony iii n-butoxide, tributoxystibine
YGBFTDQFAKDXBZ-UHFFFAOYSA-N
antimony(3+);butan-1-olate
102217
Liquid
1.264 g/mL
Moisture sensitive
2g
C12H27O3Sb
UN3082
Soluble in anhydrous butanol
CCCC[O-].CCCC[O-].CCCC[O-].[Sb+3]
341.105
341.1
≥99%

Chemical Identifiers

2155-74-0
341.105
YGBFTDQFAKDXBZ-UHFFFAOYSA-N
102217
CCCC[O-].CCCC[O-].CCCC[O-].[Sb+3]
C12H27O3Sb
MFCD00015238
1-butanol, antimony 3+ salt, acmc-20als1, antimony 3+ ; butan-1-olate, antimony 3+ ion tris butan-1-olate, antimony 3+ tributan-1-olate, antimony 3+ tris butan-1-olate, antimony iii butan-1-olate, antimony iii butoxide, antimony iii n-butoxide, tributoxystibine
antimony(3+);butan-1-olate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 218-456-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents