Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) methoxide, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Antimony (III) methoxide
CAS 29671-18-9
Molecular Formula Sb(OCH3)3
Formula Weight 214.8
MDL Number MFCD00014882
UN Number UN3181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4285304

  SDS
alfa aesar™
4285304
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4285309

  SDS
alfa aesar™
4285309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.8
Flash Point >110°C (230°F)
Chemical Name or Material Antimony (III) methoxide
Sensitivity Air & Moisture Sensitive
Melting Point 124°C
Assay 99.5%
MDL Number MFCD00014882
Molecular Formula Sb(OCH3)3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P273
Avoid release to the environment.
CAS 29671-18-9
UN Number UN3181
Quantity 2g
Assay Percent Notes (metals basis)
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Physical Form Crystalline
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Safety and Handling

Warning

GHS P Statement
P273
Avoid release to the environment.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.