Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) methoxide, 95%

Chemical Identifiers

Chemical Name or Material Antimony(III) methoxide
CAS 29671-18-9
Formula Weight 214.8
MDL Number MFCD00014882
UN Number UN3181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3349504

  SDS
alfa aesar™
3349504
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.8
Chemical Name or Material Antimony(III) methoxide
Physical Form Crystalline
Assay 95%
MDL Number MFCD00014882
CAS 29671-18-9
UN Number UN3181
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Melting Point 124°C
Health Hazard 3 GHS P Statement
P273
Avoid release to the environment.
Quantity 2g
Flash Point >110° (230°F)
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Sensitivity Air & Moisture Sensitive
Safety and Handling

Warning

GHS P Statement
P273
Avoid release to the environment.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.