missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) chloride, ultra dry, 99.999% (metals basis)

$226.54 - $2,311.43

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimonous chloride, antimontrichlorid, antimony butter, antimony chloride, antimony iii chloride, antimony trichloride, butter of antimony, caustic antimony, stibine, trichloro, trichlorostibine
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES Cl[Sb](Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4344914
View Documents
Alfa Aesar™
4344914
25g Each for $226.54
 
AA4344922
View Documents
Alfa Aesar™
4344922
100g Each for $648.45
 
AA4344936
View Documents
Alfa Aesar™
4344936
500g Each for $2,311.43
 
Description

Description

  • Ampouled under argon
Specifications

Chemical Identifiers

10025-91-9
228.11
FAPDDOBMIUGHIN-UHFFFAOYSA-K
24814
trichlorostibane
Cl3Sb
MFCD00011212
antimonous chloride, antimontrichlorid, antimony butter, antimony chloride, antimony iii chloride, antimony trichloride, butter of antimony, caustic antimony, stibine, trichloro, trichlorostibine
CHEBI:74856
Cl[Sb](Cl)Cl

Specifications

10025-91-9
MFCD00011212
14,707
It is soluble in alcohol and acetone and acids.
Cl[Sb](Cl)Cl
228.11
CHEBI:74856
Crystals,Ampouled under argon
3.14 g/mL
Ampule
223.5°C
25g
(metals basis)
Cl3Sb
UN1733
antimonous chloride, antimontrichlorid, antimony butter, antimony chloride, antimony iii chloride, antimony trichloride, butter of antimony, caustic antimony, stibine, trichloro, trichlorostibine
FAPDDOBMIUGHIN-UHFFFAOYSA-K
trichlorostibane
24814
228.11
99.999%
Pungent
Moisture sensitive
73.4°C
Antimony (III) chloride, Ultra dry
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANTIMONY TRICHLORIDE, SOLID

EINECSNumber : 233-047-2

RTECSNumber : CC4900000

TSCA : Yes

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures