missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) chloride, ultra dry, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4344922

 View more versions of this product

Catalog No. AA4344922



Description

Description

  • Ampouled under argon
Specifications

Specifications

Antimony (III) chloride
10025-91-9
Cl3Sb
UN1733
antimonous chloride, antimontrichlorid, antimony butter, antimony chloride, antimony iii chloride, antimony trichloride, butter of antimony, caustic antimony, stibine, trichloro, trichlorostibine
FAPDDOBMIUGHIN-UHFFFAOYSA-K
trichlorostibane
24814
228.11
99.999%
Pungent
Moisture sensitive
73.4°C
Ultra dry
(metals basis)
MFCD00011212
14,707
It is soluble in alcohol and acetone and acids.
Cl[Sb](Cl)Cl
228.11
CHEBI:74856
Crystals,Ampouled under argon
3.14 g/mL
Ampule
223.5°C
100g

Chemical Identifiers

10025-91-9
228.11
FAPDDOBMIUGHIN-UHFFFAOYSA-K
24814
trichlorostibane
Cl3Sb
MFCD00011212
antimonous chloride, antimontrichlorid, antimony butter, antimony chloride, antimony iii chloride, antimony trichloride, butter of antimony, caustic antimony, stibine, trichloro, trichlorostibine
CHEBI:74856
Cl[Sb](Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANTIMONY TRICHLORIDE, SOLID

EINECSNumber : 233-047-2

RTECSNumber : CC4900000

TSCA : Yes

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

Documents