missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony(III) Chloride, ACS, For Analysis, EMSURE™, MilliporeSigma™

Manufacturer:  MilliporeSigma 1078381000

 View more versions of this product

Catalog No. M1078381000



Description

Description

For analysis
Specifications

Specifications

Antimony(III) chloride
Cl3Sb
Colorless solid
FAPDDOBMIUGHIN-UHFFFAOYSA-K
trichlorostibane
24814
228.1
ACS
Glass bottle
1kg
10025-91-9
UN 1733
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
Cl[Sb](Cl)Cl
228.11
CHEBI:74856
Solid
Stinging
0.16 hPa at 20°C

Chemical Identifiers

10025-91-9
228.11
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
CHEBI:74856
Cl[Sb](Cl)Cl
Cl3Sb
FAPDDOBMIUGHIN-UHFFFAOYSA-K
24814
trichlorostibane
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.
H411: Toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 233-047-2

Documents