missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) bromide, ultra dry, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4373214

 View more versions of this product

Catalog No. AA4373214



Description

Description

  • Ampouled under argon
Specifications

Specifications

Antimony (III) bromide
7789-61-9
Br3Sb
UN3260
antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
Br[Sb](Br)Br
361.472
361.48
99.999%
Ampule
1.74
280°C
25g
Ultra dry
(metals basis)
MFCD00016317
14,706
RPJGYLSSECYURW-UHFFFAOYSA-K
tribromostibane
24615
Lumps
4.418g/mL
5 to 10mm
Air Sensitive
96.6°C

Chemical Identifiers

7789-61-9
361.472
RPJGYLSSECYURW-UHFFFAOYSA-K
24615
Br[Sb](Br)Br
Br3Sb
MFCD00016317
antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
tribromostibane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents