missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) acetate, 97%

Manufacturer:  Alfa Aesar™ 8765709

 View more versions of this product

Catalog No. AA8765709



Specifications

Specifications

Antimony (III) acetate
10g
C6H9O6Sb
UN1549
Decomposes in water
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sb+3]
298.892
298.88
97%
6923-52-0
Moisture Sensitive
MFCD00014974
antimony triacetate, antimony iii acetate, antimony acetate, acetic acid, antimony 3+ salt, octan antimonity czech, antimony 3+ triacetate, acetic acid, antimony 3+ salt 3:1, acetic acid, trianhydride with antimonic acid h3sbo3, acetic acid, antimony salt, octan antimonity
JVLRYPRBKSMEBF-UHFFFAOYSA-K
antimony(3+);triacetate
23354
Powder

Chemical Identifiers

6923-52-0
298.892
JVLRYPRBKSMEBF-UHFFFAOYSA-K
23354
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sb+3]
C6H9O6Sb
MFCD00014974
antimony triacetate, antimony iii acetate, antimony acetate, acetic acid, antimony 3+ salt, octan antimonity czech, antimony 3+ triacetate, acetic acid, antimony 3+ salt 3:1, acetic acid, trianhydride with antimonic acid h3sbo3, acetic acid, antimony salt, octan antimonity
antimony(3+);triacetate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.

TSCA : TSCA

SDS Documents