missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Antimony ICP Standard, 1000 ppm Sb in H2O/Tartaric Acid/tr HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PSB1KW100
Specifications
Antimony ICP Standard | |
Natural LDPE Bottle | |
96.0,0.72,0.57,0.14,0.1 | |
Colorless | |
Sb | |
WATWJIUSRGPENY-UHFFFAOYSA-N | |
antimony | |
5354495 | |
1 mL = 1 mg Sb |
7440-36-0,7697-37-2,7647-01-0,87-69-4,7732-18-5 | |
100 mL | |
100.0,0.75,0.59,0.15,0.1 | |
1,000ppm Sb | |
MFCD00011211 MFCD00134030 | |
[Sb] | |
121.76 | |
CHEBI:30304 | |
Liquid |
Chemical Identifiers
7440-36-0 | |
121.76 | |
WATWJIUSRGPENY-UHFFFAOYSA-N | |
CHEBI:30304 | |
[Sb] |
Sb | |
MFCD00011211 MFCD00134030 | |
5354495 | |
antimony |