missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony ICP Standard, 1000 ppm Sb in H2O/Tartaric Acid/tr HNO3, Ricca Chemical

$270.19 - $270.19

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
PubChem CID 5354495
ChEBI CHEBI:30304
IUPAC Name antimony
SMILES [Sb]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PSB1KW-100
View Documents Promotion Details
Ricca Chemical Company
PSB1KW100
100 mL Natural LDPE Bottle
Each for $270.19
 
Description

Description

Specifications

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
CHEBI:30304
[Sb]
Sb
MFCD00011211 MFCD00134030
5354495
antimony

Specifications

7440-36-0,7697-37-2,7647-01-0,87-69-4,7732-18-5
100 mL
100.0,0.75,0.59,0.15,0.1
1,000ppm Sb
MFCD00011211 MFCD00134030
[Sb]
121.76
CHEBI:30304
Liquid
Natural LDPE Bottle
96.0,0.72,0.57,0.14,0.1
Colorless
Sb
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
5354495
1 mL = 1 mg Sb
Antimony ICP Standard
SDS
missing translation for 'documents'

missing translation for 'documents'