missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony Broken rod, 12.7mm (0.50 in.) dia x various lengths, 99.8% (Metals basis), Alfa Aesar™

Form: Broken pieces from cast rod

Manufacturer:  Alfa Aesar 45047A3

 View more versions of this product

Catalog No. AA45047A3



Specifications

Specifications

Antimony
99.8% (Metals basis)
MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
[Sb]
121.76
CHEBI:30304
Odorless
2kg
7440-36-0
Sb
Silver Gray rod
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
5354495
Broken Rod, 12.7mm (0.50 in.) dia X various lengths
Silver to Gray

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
5354495
antimony
Sb
MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
CHEBI:30304
[Sb]
SDS Documents