Please login to your online account to display your discounted pricing

Antimony AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Sb), LabChem™

$43.00 - $92.51

Chemical Identifiers

Chemical Name or Material Antimony AA Standard
CAS 7732-18-5
CAS Max % 93.01
CAS 7647-01-0
CAS Max % 6.8
CAS 10025-91-9
CAS Max % 0.19
Name Note 1000ppm (1mL = 1mg Sb)
Purity Grade Certified Grade
Linear Formula SbCl3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC114201

 
lab chem
LC114201
Poly Bottle 500mL Each for $92.51

LC114207

 
lab chem
LC114207
Poly Bottle 125mL Each for $43.00
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 228.11
Chemical Name or Material Antimony AA Standard
Specific Gravity 1g/mL
Color Clear, colorless
Quantity 500mL
Decomposition Information Hydrogen chloride; Antimony and its oxides; Thermal decomposition generates: Corrosive vapors
CAS Max % 0.19
Name Note 1000ppm (1mL = 1mg Sb)
Physical Form Liquid
Concentration 1000ppm ±10ppm (1mL = 1mg Sb)
Linear Formula SbCl3
CAS 7732-18-5
UN Number UN3264
Health Hazard 1 Danger
Identification Passes Test
CAS Max % 93.01
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS 7647-01-0
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS Max % 6.8
Recommended Storage Room Temperature
Density 1g/mL
CAS 10025-91-9
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.

Room Temperature