Learn More
Antimony AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Sb), LabChem™
Supplier: LabChem LC114201
Specifications
Antimony AA Standard | |
1000ppm (1mL = 1mg Sb) | |
Poly Bottle | |
93.01,6.8,0.19 | |
Sb | |
MFCD00011211 MFCD00134030 | |
WATWJIUSRGPENY-UHFFFAOYSA-N | |
antimony | |
5354495 | |
228.11 | |
Passes Test | |
Soluble in water |
1000ppm ±10ppm (1mL = 1mg Sb) | |
7440-36-0,7647-01-0,10025-91-9,7732-18-5 | |
500 mL | |
Colorless | |
SbCl3 | |
UN3264 | |
[Sb] | |
121.76 | |
CHEBI:30304 | |
Certified | |
1g/mL | |
Liquid |
Chemical Identifiers
7440-36-0 | |
121.76 | |
WATWJIUSRGPENY-UHFFFAOYSA-N | |
CHEBI:30304 | |
[Sb] |
Sb | |
MFCD00011211 MFCD00134030 | |
5354495 | |
antimony |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature