Please login to your online account to display your discounted pricing

Alfa Aesar™ Antileukinate

For cellular and molecular biology applications

$200.41 - $307.38

Specifications

Product Type Antileukinate
Molecular Weight 1003.20
Storage Requirements -30° to -10°C
Format Solid
Sequence N-Acetyl-Arg-Arg-Trp-Trp-Cys-Arg-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66875EXD

 
alfa aesar™
J66875EXD
0.5mg Each for $200.41

AAJ66875MCR

 
alfa aesar™
J66875MCR
1mg Each for $307.38
Description & Specifications

Specifications

Product Type Antileukinate
Molecular Weight 1003.20
Storage Requirements -30° to -10°C
Format Solid
Sequence N-Acetyl-Arg-Arg-Trp-Trp-Cys-Arg-NH2
Regulatory Status RUO
Purification Method HPLC