Please login to your online account to display your discounted pricing

Alfa Aesar™ Antileukinate


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66875MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66875MCR

  • / Each
 

Description & Specifications

Specifications

Quantity 1mg
Format Solid
Sequence N-Acetyl-Arg-Arg-Trp-Trp-Cys-Arg-NH2
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Antileukinate
Molecular Weight 1003.20
Purification Method HPLC