missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antiestrogen

$150.54 - $212.42

Chemical Identifiers

CAS 166664-90-0
Molecular Formula C64H104N13O22PS
Formula Weight 1470.64
MDL Number MFCD03787962
Synonym Cys-Asn-Val-Val-Pro-Leu-Tyr(PO{3}H2)-Asp-Leu-Leu-Leu-Glu
Chemical Name or Material Antiestrogen
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66374EXD Alfa Aesar™
J66374EXD
0.5mg Each for $150.54
AAJ66374MCR Alfa Aesar™
J66374MCR
1mg Each for $212.42
Specifications Description & Specifications

Chemical Identifiers

166664-90-0
1470.64
Cys-Asn-Val-Val-Pro-Leu-Tyr(PO{3}H2)-Asp-Leu-Leu-Leu-Glu
C64H104N13O22PS
MFCD03787962
Antiestrogen

Specifications

166664-90-0
C64H104N13O22PS
MFCD03787962
White to off-white
Antiestrogen
Solid
1470.64
Cys-Asn-Val-Val-Pro-Leu-Tyr(PO{3}H2)-Asp-Leu-Leu-Leu-Glu
0.5mg

Safety and Handling

storageNote1 : -30° to -10°C

Recommended Storage : −30° to −10°C