Antibiotic Sensitivity Discs
Study antibiotic sensitivity and resistance.
Manufacturer:  Fisher Scientific B31264
 View more versions of this product
Catalog No. S70151A
Description
Description
  • Vials are sealed for storage
  • Designed to dispense one disc at a time, without contact or spoilage of remaining discs
Order Info
Vials and bottles are supplied in packs of 50.
Specifications
Specifications
Education
Disc
10μg
Ampicillin
50 Disks
Documents