missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
zinc finger, SWIM-type containing 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ZSWIM2 protein.
Supplier: Abnova Corporation H00151112B01P
Description
Sequence: MLRRGYKASERRRHLSERLSWHQDQALSSSIYLLREMGPTGFLLREEEPEYMDFRVFLGNPHVCNCSTFPKGGELCKHICWVLLKKFELPRNHESALQLGLGEREISDLLRGIHRVQTPQPGTNDENEHVEEDGYIKQKEIDSEDICSICQELLLEKKLPVTFCRFGCGNSIHIKCMKILANYQSTSNTSMLKCPLCRKEFAPLKLILEEFKNSSKLVAAAEKERLDKHLGIPCNNCKQFPIEGKCYKCTECIEYHLCQECFDSYCHLSHTFTFREKRNQKWRSLEKRADEVVKYIDTKNETEEKMSHFQEKQGQVYTPKHIVRSLPLQLITKNSKLLAPGYQCLLCLKAFHLGQHTRLLPCTHKFHRKCIDNWLFHKCNSCPIGGQVIYNPLTWKNSAVNGQAHQSVSNRDIIHLSKQKEPDLFIPGTGLVLKQNRLGILPSIPQCNFDELNTPQSPKDAYENTTIDNLCSIKLDNSNSKKLTYEYKISQHFPRYLQDLPTVSFGKIPSQTLLPPIVHKNIVCPTAMESPCISGKYHTSLSRVTKGCKCNNHNLKKTPATKIREDNKRSTLLPEDFNLIVNWSTAKLSLSKRYSNCMGEITRKCSHLSRQPVSHSVNTKSTELSLIIEGVQLSpecifications
zinc finger, SWIM-type containing 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ZSWIM2 protein. | |
ZSWIM2 | |
MGC33890/ZZZ2 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC031094.1 | |
ZSWIM2 | |
ZSWIM2 (AAH31094.1, 1 a.a. ∼ 633 a.a) full-length human protein. | |
50 μg | |
Ubiquitin/Proteasome | |
151112 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |