Please login to your online account to display your discounted pricing

anti-ZNF521, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP191270

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP191270

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q96K83
Concentration 1.07mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Host Species Rabbit
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene ID (Entrez) 25925
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen A synthetic peptide made to an internal portion of the human ZNF521 protein (between residues 200-250) [UniProt Q96K83]
Antigen ZNF521