missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-ZNF521, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human ZNF521 protein (between residues 200-250) [UniProt Q96K83]
Antigen ZNF521
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP191270 Novus Biologicals Canada
NBP191270
0.1mL Each for $309.05
NBP191270SS Novus Biologicals Canada
NBP191270SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
ZNF521
Affinity Purified
RUO
Q96K83
PBS, 30% glycerol
A synthetic peptide made to an internal portion of the human ZNF521 protein (between residues 200-250) [UniProt Q96K83]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin)
25925
1.07mg/mL