Please login to your online account to display your discounted pricing

anti-ZNF521, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP191271

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP191271

  • / Each
 

Description & Specifications

Specifications

Immunogen A synthetic peptide made to an internal portion of the human ZNF521 protein (between residues 450-550) [UniProt Q96K83]
Gene ID (Entrez) 25925
Concentration 1.11mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Antigen ZNF521
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q96K83
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled