Please login to your online account to display your discounted pricing

anti-ZNF521, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP191270SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP191270SS

  • / Each
 

Description & Specifications

Specifications

Antigen ZNF521
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1.07mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q96K83
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human ZNF521 protein (between residues 200-250) [UniProt Q96K83]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 25925
Target Species Human
Target Species Murine