missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
zinc finger protein 266, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ZNF266 protein.
Supplier: Abnova Corporation H00010781B01
Description
Sequence: MKIHVGEKPYECKECGIAFTRSSQLTEHLKTHTAKDPFECKICGKSFRNSSCLSDHFRIHTGIKPYKCKDCGKAFTQNSDLTKHARTHSGERPYECKECGKAFARSSRLSEHTRTHTGEKPFECVKCGKAFAISSNLSGHLRIHTGEKPFECLECGKAFTHSSSLNNHMRTHSAKKPFTCMECGKAFKFPTCVNLHMRIHTGEKPYKCKQCGKSFSYSNSFQLHERTHTGEKPYECKECGKAFSSSSSFRNHERRHADERLSASpecifications
zinc finger protein 266 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ZNF266 protein. | |
ZNF266 | |
HZF1 | |
Mouse | |
50 μL | |
Yes | |
10781 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
BC017407.1 | |
ZNF266 | |
ZNF266 (AAH17407.1, 1 a.a. ∼ 263 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |