Please login to your online account to display your discounted pricing

anti-ZMPSTE24, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human ZMPSTE24 protein sequence (between residues 1-100). [Swiss-Prot O75844]
Antigen ZMPSTE24
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002387

 
novus biologicals canada
NB1002387
Unlabeled 0.1mL Each for $329.00

NB1002387SS

 
novus biologicals canada
NB1002387SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human ZMPSTE24 protein sequence (between residues 1-100). [Swiss-Prot O75844]
Antigen ZMPSTE24
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human, mouse and zebrafish. Predicted to react with primate based on 100% sequence homology.
Gene Accession No. O75844
Gene ID (Entrez) 10269
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL