Please login to your online account to display your discounted pricing

anti-ZMPSTE24, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002387SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002387SS

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Formulation Tris-glycine, 150mM NaCl
Primary or Secondary Primary
Gene Accession No. O75844
Gene ID (Entrez) 10269
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Zebrafish
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.025mL
Antigen ZMPSTE24
Test Specificity Human, mouse and zebrafish. Predicted to react with primate based on 100% sequence homology.
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Format Affinity Purified
Immunogen A synthetic peptide made to an N-terminal portion of the human ZMPSTE24 protein sequence (between residues 1-100). [Swiss-Prot O75844]
Host Species Rabbit