Please login to your online account to display your discounted pricing

anti-ZMPSTE24, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002387

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002387

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10269
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Zebrafish
Host Species Rabbit
Formulation Tris-glycine, 150mM NaCl
Test Specificity Human, mouse and zebrafish. Predicted to react with primate based on 100% sequence homology.
Primary or Secondary Primary
Gene Accession No. O75844
Antigen ZMPSTE24
Format Affinity Purified
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen A synthetic peptide made to an N-terminal portion of the human ZMPSTE24 protein sequence (between residues 1-100). [Swiss-Prot O75844]
Quantity 0.1mL