Please login to your online account to display your discounted pricing

anti-ZMPSTE24, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002388SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002388SS

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-glycine, 150mM NaCl
Gene Accession No. O75844
Primary or Secondary Primary
Concentration 1.0mg/mL
Antigen ZMPSTE24
Format Affinity Purified
Host Species Rabbit
Applications Western Blotting
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10269
Conjugate Unlabeled
Immunogen A synthetic peptide made to a C-terminal portion of the human ZMPSTE24 protein sequence (between residues 400-475). [Swiss-Prot O75844]
Regulatory Status RUO
Cross Reactivity Human