Please login to your online account to display your discounted pricing

anti-Zic2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 350 to 400 of human ZIC2 was used as immunogen.
Antigen Zic2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224607SS

 
novus biologicals canada
NBP224607SS
Unlabeled 0.025mg Each for $109.00

NBP224607

 
novus biologicals canada
NBP224607
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 350 to 400 of human ZIC2 was used as immunogen.
Antigen Zic2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, mouse and rat, 94% homologous in cow, 88% homologous in xenopus and chicken, and 75% homologous in pufferfish.
Gene Accession No. O95409
Gene ID (Entrez) 7546
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL