Please login to your online account to display your discounted pricing

anti-ZIC1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Zic1 protein (between residues 20-75) [UniProt Q15915]
Antigen ZIC1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600488

 
novus biologicals canada
NB600488
Unlabeled 0.1mL Each for $329.00

NB600488SS

 
novus biologicals canada
NB600488SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Zic1 protein (between residues 20-75) [UniProt Q15915]
Antigen ZIC1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human and mouse. Predicted to react with rat based on 100% sequence homology.
Gene Accession No. Q15915
Gene ID (Entrez) 7545
Formulation PBS, 30% glycerol
Concentration 1.0mg/mL