Please login to your online account to display your discounted pricing

Zfp110 Polyclonal Antibody, Invitrogen™


Zfp110 Antibody, OSZ00001W, from Invitrogen™. Species Reactivity: Mouse, Rat; Applications: Immunohistochemistry.

Manufacturer: pierce biotechnology inc  OSZ00001W

Catalog No. PIOSZ00001W

  • / Each
 

Description & Specifications

Specifications

Antigen Zfp110
Applications Immunohistochemistry
Conjugate Unlabeled
Format Serum
Formulation 100μL unpurified antibody in whole serum and no preservative
Gene Accession No. Q4V8E9, Q923B3
Gene Alias Zfp110
Gene Symbols NRIF1
Host Species Rabbit
Immunogen A synthetic peptide from c-terminal region of rat zinc finger protein 110
Quantity 100μL
Storage Requirements avoiding freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 308362, 65020
Target Species Murine
Target Species Rat

Zfp110 Antibody, OSZ00001W, from Invitrogen. Species Reactivity: Mouse, Rat; Applications: Immunohistochemistry.


Host Species: Rabbit
Cross Reactivity: Mouse, Rat
Immunogen: A synthetic peptide from c-terminal region of rat zinc finger protein 110
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Zfp110
Uniprot Accession: Q4V8E9, Q923B3
Entrez GeneID: 308362, 65020
Gene Symbol: NRIF1
Reconstitute with 100 ul of distilled water.