Please login to your online account to display your discounted pricing

anti-Zfp110 Polyclonal, Thermo Scientific™

Zfp110 Polyclonal antibody OSZ00001W from Thermo Scientific™ specifically detects Zfp110 in Mouse and Rat samples, and it is validated for Immunohistochemistry

$448.05 - $448.05

Specifications

Antigen Zfp110
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSZ00001W

 
pierce biotechnology inc
OSZ00001W
Unlabeled 100μL Each for $448.05

PIOSZ00002G

 
pierce biotechnology inc
OSZ00002G
Unlabeled 500μg Each for $448.05
Description & Specifications

Specifications

Antigen Zfp110
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit
Immunogen A synthetic peptide from c-terminal region of rat zinc finger protein 110
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Zfp110 Polyclonal antibody OSZ00001W from Thermo Scientific™ specifically detects Zfp110 in Mouse and Rat samples, and it is validated for Immunohistochemistry


Host Species: Rabbit
Cross Reactivity: Mouse, Rat
Immunogen: A synthetic peptide from c-terminal region of rat zinc finger protein 110
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Zfp110
Uniprot Accession: Q4V8E9, Q923B3
Gene Symbol: Nrif1
Entrez GeneID: 308362, 65020
OSZ00001W detects Zfp110 in Mouse and Rat samples. OSZ00001W has been successfully used in Immunohistochemistry procedures. OSZ00001W The immunogen is A synthetic peptide from c-terminal region of rat zinc finger protein 110.