Please login to your online account to display your discounted pricing

Zfp110 Polyclonal Antibody, Invitrogen™


Zfp110 Antibody, OSZ00002G, from Invitrogen™. Species Reactivity: Mouse, Rat; Applications: Immunohistochemistry.

Manufacturer: pierce biotechnology inc  OSZ00002G

Catalog No. PIOSZ00002G

  • / Each
 

Description & Specifications

Specifications

Antigen Zfp110
Applications Immunohistochemistry
Conjugate Unlabeled
Format Purified
Formulation 500μg purified antibody (1mg/mL) and no preservative
Gene Accession No. Q4V8E9, Q923B3
Gene Alias Zfp110
Gene Symbols NRIF1
Host Species Rabbit
Immunogen A synthetic peptide from c-terminal region of rat zinc finger protein 110
Isotype IgG
Quantity 500μg
Storage Requirements avoiding freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 308362, 65020
Target Species Murine
Target Species Rat

Zfp110 Antibody, OSZ00002G, from Invitrogen. Species Reactivity: Mouse, Rat; Applications: Immunohistochemistry.


Host Species: Rabbit
Cross Reactivity: Mouse, Rat
Isotype: IgG
Immunogen: A synthetic peptide from c-terminal region of rat zinc finger protein 110
Formulation: 500μg purified antibody (1mg/mL) and no preservative
Synonyms: Zfp110
Uniprot Accession: Q4V8E9, Q923B3
Entrez GeneID: 308362, 65020
Gene Symbol: NRIF1
Reconstitute with 500 ul of distilled water.