missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-ZEB1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an C-terminal portion of the human ZEB1 protein (between residues 1087-1124). [UniProt P37275]
Antigen ZEB1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP213159SS Novus Biologicals Canada
NBP213159SS
0.025mL Each for $139.00
NBP213159 Novus Biologicals Canada
NBP213159
0.1mL Each for $309.05
Specifications Description & Specifications

Specifications

Rabbit
ZEB1
Affinity Purified
RUO
P37275
PBS, 30% glycerol
A synthetic peptide made to an C-terminal portion of the human ZEB1 protein (between residues 1087-1124). [UniProt P37275]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Simple Western,Western Blot
6935
1.00mg/mL