Please login to your online account to display your discounted pricing

anti-ZBTB7A/Pokemon, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 1-50 of human Pokemon was used as the immunogen for this antibody.
Antigen ZBTB7A/Pokemon
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223673SS

 
novus biologicals canada
NBP223673SS
Unlabeled 0.025mL Each for $109.00

NBP223673

 
novus biologicals canada
NBP223673
Unlabeled 0.1mL Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 1-50 of human Pokemon was used as the immunogen for this antibody.
Antigen ZBTB7A/Pokemon
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 95% homologous in mouse, rat, cow, horse and opossum, 86% homologous in dog and 77% homologous in chicken.
Gene ID (Entrez) 51341
Formulation 100μL of crude rabbit sera with 50% glycerol