missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-ZBTB2, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 450 to 500 of human ZBTB2 was used as immunogen for the antibody.
Antigen ZBTB2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224660 Novus Biologicals Canada
NBP224660
0.1mg Each for $257.41
NBP224660SS Novus Biologicals Canada
NBP224660SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
ZBTB2
Purified
RUO
The amino acid sequence used as immunogen for this antibody is 100% homologous in human (isoform CRA_a and CRA_b) and chimpanzee, 88% homologous in horse and cow, 82% homologous in rat and 76% homologous in mouse.
57621
0.5mg/mL
A portion of amino acids 450 to 500 of human ZBTB2 was used as immunogen for the antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
Q8N680
100μg in 200μL PBS containing 0.05% BSA