missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Ymer, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 450-500 of human Chromosome 3 open reading frame 6 (YMER) was used as the immunogen.
Antigen Ymer
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224483 Novus Biologicals Canada
NBP224483
0.1mg Each for $255.80
NBP224483SS Novus Biologicals Canada
NBP224483SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Ymer
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a) and chimpanzee, 93% homologous in rhesus monkey and 75% homologous in cow and horse.
152137
0.5mg/mL
A portion of amino acids 450-500 of human Chromosome 3 open reading frame 6 (YMER) was used as the immunogen.
Polyclonal
Primary
Western Blot
Q8IVM0
100μg in 200μL PBS containing 0.05% BSA