Please login to your online account to display your discounted pricing

anti-YB1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A C-terminal peptide of human YB1. [Swiss-Prot No. P67809]
Antigen YB1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP197572

 
novus biologicals canada
NBP197572
Unlabeled 0.1mL Each for $309.05

NBP197572SS

 
novus biologicals canada
NBP197572SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A C-terminal peptide of human YB1. [Swiss-Prot No. P67809]
Antigen YB1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Test Specificity Human, Mouse and Rat. Based on immunogen's sequence homology, this antibody is predicted to react with Bovine (100%), Rabbit (100%), Chicken (93%) and Xenopus (87%).
Gene Accession No. P67809
Gene ID (Entrez) 4904