Please login to your online account to display your discounted pricing

anti-YB1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197572SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP197572SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene Accession No. P67809
Test Specificity Human, Mouse and Rat. Based on immunogen's sequence homology, this antibody is predicted to react with Bovine (100%), Rabbit (100%), Chicken (93%) and Xenopus (87%).
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Immunogen A C-terminal peptide of human YB1. [Swiss-Prot No. P67809]
Quantity 0.025mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4904
Primary or Secondary Primary
Format Antiserum
Regulatory Status RUO
Host Species Rabbit
Antigen YB1