missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-YAP1, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen Recombinant human YAP1 protein expressed in bacteria. [UniProt No. P46937]
Antigen YAP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB11058358T Novus Biologicals Canada
NB11058358SS
0.025mL Each for $144.63
NB11058358 Novus Biologicals Canada
NB11058358
0.1mL Each for $384.00
Specifications Description & Specifications

Specifications

Rabbit
YAP1
Affinity Purified
RUO
P46937
PBS, 15% glycerol
Recombinant human YAP1 protein expressed in bacteria. [UniProt No. P46937]
Polyclonal
Primary
Chromatin Immunoprecipitation,Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunoprecipitation,Simple Western,Western Blot
10413
0.2mg/mL