Please login to your online account to display your discounted pricing

anti-XRCC3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100165SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100165SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Applications Immunoprecipitation
Applications Western Blotting
Concentration 1mg/mL
Gene Accession No. O43542
Primary or Secondary Primary
Gene ID (Entrez) 7517
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Antigen XRCC3
Format Affinity Purified
Host Species Rabbit
Immunogen A synthetic partial peptide of human XRCC3. [UniProt No. O43542]
Regulatory Status RUO
Cross Reactivity Human