Please login to your online account to display your discounted pricing

anti-XRCC3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100165

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100165

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O43542
Gene ID (Entrez) 7517
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Immunogen A synthetic partial peptide of human XRCC3. [UniProt No. O43542]
Antigen XRCC3
Conjugate Unlabeled
Concentration 1mg/mL
Formulation PBS
Primary or Secondary Primary
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Host Species Rabbit
Quantity 0.1mL