Please login to your online account to display your discounted pricing

anti-XIAP, Polyclonal, Novus Biologicals™

Manufacturer: novus biologicals canada  NB10056186

 View all options for this product


For Research Use Only

Catalog No. NB10056186


Description & Specifications


Antigen XIAP
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antiserum
Formulation PBS
Host Species Rabbit
Immunogen 1999).
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 331
Test Specificity The antibody recognizes epitopes in the BIR2 domain of XIAP. Therefore it can recognize full-length XIAP and XIAP cleavage fragments containing the BIR2 domain. However, XIAP cleavage fragments may be biologically unstable, and therefore cleavage fragments may be difficult to detect. Cross reacts with Gerbil, Human, Mouse and Rat.