Please login to your online account to display your discounted pricing

anti-XIAP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056184SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056184T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 331
Test Specificity The antibody recognizes epitopes in the BIR2 domain of XIAP. Therefore it can recognize full-length XIAP and XIAP cleavage fragments containing the BIR2 domain. However, XIAP cleavage fragments may be biologically unstable, and therefore cleavage fragments may be difficult to detect. Cross reacts with Gerbil, Human, Mouse and Rat.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen 1999).
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Monoclonal or Polyclonal Polyclonal
Formulation PBS
Primary or Secondary Primary
Format Antiserum
Host Species Rabbit
Quantity 0.025mL
Antigen XIAP