Please login to your online account to display your discounted pricing

anti-XIAP, Polyclonal, Novus Biologicals™

$160.00 - $246.00

Specifications

Host Species Rabbit
Immunogen 1999).
Antigen XIAP
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056186

 
novus biologicals canada
NB10056186
Unlabeled 0.05mL Each for $246.00

NB10056186T

 
novus biologicals canada
NB10056186SS
Unlabeled 0.025mL Each for $160.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 1999).
Antigen XIAP
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity The antibody recognizes epitopes in the BIR2 domain of XIAP. Therefore it can recognize full-length XIAP and XIAP cleavage fragments containing the BIR2 domain. However, XIAP cleavage fragments may be biologically unstable, and therefore cleavage fragments may be difficult to detect. Cross reacts with Gerbil, Human, Mouse and Rat.
Gene ID (Entrez) 331
Formulation PBS