Please login to your online account to display your discounted pricing

anti-XBP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177681SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177681SS

  • / Each
 

Description & Specifications

Specifications

Antigen XBP1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 0.58mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P17861
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human XBP1 protein (within residues 100-250). [Swiss-Prot P17861]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7494
Target Species Human
Target Species Murine